missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CFTR Rabbit anti-Human, Clone: CFTR/7003R, Novus Biologicals™
Rabbit Monoclonal Antibody
£238.00 - £429.00
Specifications
| Antigen | CFTR |
|---|---|
| Clone | CFTR/7003R |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18301507
|
Novus Biologicals
NBP3-13843-20UG |
20 μg |
£238.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18361867
|
Novus Biologicals
NBP3-13843-100UG |
100 μg |
£429.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CFTR Monoclonal antibody specifically detects CFTR in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| CFTR | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| ABC Transporters, Cancer, Diabetes Research, Lipid and Metabolism, Membrane Trafficking and Chaperones | |
| 10 mM PBS with 0.05% BSA | |
| ABC35, ABCC7CF, ATP-binding cassette sub-family C member 7, ATP-binding cassette transporter sub-family C member 7, cAMP-dependent chloride channel, CFTR/MRP, Channel conductance-controlling ATPase, cystic fibrosis transmembrane conductance regulator, cystic fibrosis transmembrane conductance regulator (ATP-binding cassettesub-family C, member 7), cystic fibrosis transmembrane conductance regulator, ATP-binding cassette(sub-family C, member 7), dJ760C5.1, EC 3.6.3, MRP7EC 3.6.3.49, TNR-CFTR | |
| Recombinant fragment (around aa550-850) of human CFTR protein (exact sequence is proprietary) (Uniprot: P13569 ) | |
| Primary | |
| Store at 4°C. |
| CFTR/7003R | |
| Immunohistochemistry-Paraffin 1-2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P13569 | |
| 1080 | |
| IgG κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title