missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CFTR Mouse, Clone: CFTR/1643, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 1080-MSM3-P0
This item is not returnable.
View return policy
Specifications
| CFTR | |
| Cell Surface and Cytoplasmic | |
| CFTR/1643 | |
| Unconjugated | |
| NP_011514056 | |
| Mouse | |
| 165-170kDa | |
| Protein A/G Purified | |
| RUO | |
| 1080 | |
| IgG2b |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% Azide | |
| CFTR | |
| Recombinant human CFTR fragment (aa258-385) | |
| MOLT-4 cells. Pancreas, Kidney or Placenta. | |
| 20 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction