missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CFTR Mouse anti-Human, Mouse, Clone: rCFTR/1342, Novus Biologicals™
Mouse Monoclonal Antibody
£387.00
Specifications
| Antigen | CFTR |
|---|---|
| Clone | rCFTR/1342 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 1 to 2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
Description
CFTR Monoclonal specifically detects CFTR in Human, Mouse samples. It is validated for Immunohistochemistry-Paraffin.Specifications
| CFTR | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| ABC Transporters, Cancer, Diabetes Research, Lipid and Metabolism, Membrane Trafficking and Chaperones | |
| 10 mM PBS with 0.05% BSA | |
| 1080 | |
| IgG1 κ | |
| Protein A or G purified |
| rCFTR/1342 | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| ABC35, ABCC7CF, ATP-binding cassette sub-family C member 7, ATP-binding cassette transporter sub-family C member 7, cAMP-dependent chloride channel, CFTR/MRP, Channel conductance-controlling ATPase, cystic fibrosis transmembrane conductance regulator, cystic fibrosis transmembrane conductance regulator (ATP-binding cassettesub-family C, member 7), cystic fibrosis transmembrane conductance regulator, ATP-binding cassette(sub-family C, member 7), dJ760C5.1, EC 3.6.3, MRP7EC 3.6.3.49, TNR-CFTR | |
| Recombinant full-length human CFTR protein. | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title