missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CFTR Mouse anti-Human, Clone: CFTR/1341, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB14727.100ug
This item is not returnable.
View return policy
Description
Mouse monoclonal antibody raised against full length recombinant human CFTR.
- High specificity
- High reactivity
Specifications
| CFTR | |
| Monoclonal | |
| Unconjugated | |
| Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) Western Blotting (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| P13569 | |
| CFTR | |
| Recombinant protein corresponding to full length human CFTR. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| CFTR/1341 | |
| Mouse monoclonal antibody raised against full length recombinant human CFTR. | |
| In 10 mM PBS. | |
| ABC35/ABCC7/CF/CFTR/MRP/MRP7/TNR-CFTR/dJ760C5.1 | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| 1080 | |
| Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction