missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CFTR Monoclonal Antibody (M3A7), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA511768
This item is not returnable.
View return policy
Description
MA5-11768 targets Cystic Fibrosis Transmembrane Regulator in IF, IHC (P), IP, and WB applications and shows reactivity with Human, mouse, and Rat samples. The MA5-11768 immunogen is recombinant protein encoding NBF2 domain of human CFTR.
Cystic Fibrosis (CF) is a common lethal genetic disease caused by mutations of the gene coding for the cystic fibrosis transmembrane conductance factor, a cAMP regulated chloride channel. Approximately 70% of all CF cases share the deletion of a phenylalanine at position 508 (delta F508) which results in abnormal chloride transport. Since the CF mutation is lethal, most often by lung and liver disease, it raises the question of why this genetic disease remains as common as it is. One possible explanation is that Salmonella typhi has been shown to use CFTR to enter intestinal epithelial cells and that delta F508 heterozygote and homozygote mice showed 86% and 100% reductions in S. typhi intestinal submucosal uptake.
Specifications
| CFTR | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
| P13569, P26361, P34158 | |
| CFTR | |
| Recombinant protein encoding NBF2 domain of human CFTR | |
| 500 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| M3A7 | |
| Unconjugated | |
| CFTR | |
| ABC35; Abcc7; ATP Binding Cassette Superfamily C Member 7 (ABCC7); ATP-binding cassette sub-family C member 7; ATP-binding cassette transporter sub-family C member 7; ATP-binding cassette, subfamily c, member 7; AW495489; cAMP-dependent chloride channel; CF; CFTR; CFTR/MRP; Channel conductance-controlling ATPase; cystic fibrosis transmembrane conductance regulator; cystic fibrosis transmembrane conductance regulator (ATP-binding cassette sub-family C, member 7); cystic fibrosis transmembrane conductance regulator homolog; cystic fibrosis transmembrane conductance regulator homolog; ATP-binding cassette, subfamily c, member 7; dJ760C5.1; MRP7; RGD1561193; tcag7.78; TNR CFTR; TNR-CFTR | |
| Mouse | |
| Protein G | |
| RUO | |
| 1080, 12638, 24255 | |
| 4° C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction