missing translation for 'onlineSavingsMsg'
Learn More

CFTR Antibody (MM13-4), Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18404698 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18404698 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18404698 Supplier Novus Biologicals Supplier No. NBP229828AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CFTR Monoclonal antibody specifically detects CFTR in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen CFTR
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone MM13-4
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias ABC35, ABCC7CF, ATP-binding cassette sub-family C member 7, ATP-binding cassette transporter sub-family C member 7, cAMP-dependent chloride channel, CFTR/MRP, Channel conductance-controlling ATPase, cystic fibrosis transmembrane conductance regulator, cystic fibrosis transmembrane conductance regulator (ATP-binding cassettesub-family C, member 7), cystic fibrosis transmembrane conductance regulator, ATP-binding cassette(sub-family C, member 7), dJ760C5.1, EC 3.6.3, MRP7EC 3.6.3.49, TNR-CFTR
Host Species Mouse
Immunogen Synthetic peptide (RKGYRQRLELSD) corresponding to residues 25-36 of human cystic fibrosis transmembrane conductance regulator (CFTR).
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline ABC Transporters, Cancer, Diabetes Research, Lipid and Metabolism, Membrane Trafficking and Chaperones
Primary or Secondary Primary
Gene ID (Entrez) 1080
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.