missing translation for 'onlineSavingsMsg'
Learn More

CFTR Antibody (CFTR/7003R) - Azide and BSA Free, Novus Biologicals™

Product Code. 18331442
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18331442 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18331442 Supplier Bio-Techne Supplier No. NBP314272

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

CFTR Monoclonal antibody specifically detects CFTR in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CFTR
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CFTR/7003R
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Formulation 10mM PBS
Gene Alias ABC35, ABCC7CF, ATP-binding cassette sub-family C member 7, ATP-binding cassette transporter sub-family C member 7, cAMP-dependent chloride channel, CFTR/MRP, Channel conductance-controlling ATPase, cystic fibrosis transmembrane conductance regulator, cystic fibrosis transmembrane conductance regulator (ATP-binding cassettesub-family C, member 7), cystic fibrosis transmembrane conductance regulator, ATP-binding cassette(sub-family C, member 7), dJ760C5.1, EC 3.6.3, MRP7EC 3.6.3.49, TNR-CFTR
Host Species Rabbit
Immunogen Recombinant fragment (around aa550-850) of human CFTR protein (exact sequence is proprietary) (Uniprot: P13569 )
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline ABC Transporters, Cancer, Diabetes Research, Lipid and Metabolism, Membrane Trafficking and Chaperones
Primary or Secondary Primary
Gene ID (Entrez) 1080
Target Species Human
Content And Storage Store at -20 to -80°C. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.