missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CFTR Antibody (CFTR/1341), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-53369-100UG
This item is not returnable.
View return policy
Description
CFTR Monoclonal specifically detects CFTR in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| CFTR | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 0.5 - 1 ug/ml, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, SDS-Page | |
| ABC35, ABCC7CF, ATP-binding cassette sub-family C member 7, ATP-binding cassette transporter sub-family C member 7, cAMP-dependent chloride channel, CFTR/MRP, Channel conductance-controlling ATPase, cystic fibrosis transmembrane conductance regulator, cystic fibrosis transmembrane conductance regulator (ATP-binding cassettesub-family C, member 7), cystic fibrosis transmembrane conductance regulator, ATP-binding cassette(sub-family C, member 7), dJ760C5.1, EC 3.6.3, MRP7EC 3.6.3.49, TNR-CFTR | |
| Mouse | |
| 167 kDa | |
| 100 μg | |
| ABC Transporters, Cancer, Diabetes Research, Lipid and Metabolism, Membrane Trafficking and Chaperones | |
| 1080 | |
| Store at 4C. | |
| IgG1 |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), SDS-Page | |
| CFTR/1341 | |
| Unconjugated | |
| 10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
| CFTR | |
| Recombinant human full-length CFTR | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction