missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Cfr42I (SacII) (10 U/μL)

Product Code. 10568180 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1200 U
2,000 units
5 x 1200 units
Unit Size:
1200 units
2000 units
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10568180 1200 U 1200 units
10369679 5 x 1200 units Pack of 5
10379679 2,000 units 2000 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10568180 Supplier Thermo Scientific™ Supplier No. ER0201

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

The Cfr42I (SacII) restriction enzyme recognizes CCGC^GG sites and cuts best at 37°C in B buffer (Isoschizomers: KspI, SacII, Sfr303I, SgrBI, SstII).

Conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

Highlights

  • Superior quality – stringent quality control and industry leading manufacturing process
  • Convenient color-coded Five Buffer System
  • Includes universal Tango buffer for double-digestions
  • BSA premixed in reaction buffers
  • Wide selection of restriction endonuclease specificities

Applications

  • Molecular cloning
  • Restriction site mapping
  • Genotyping
  • Southern Blot
  • Restriction fragment length polymorphism (RFLP)
  • SNP

Note

Cfr42I activity is affected by high salt concentration. Trace amounts of sodium chloride remaining in the substrate DNA after completion of upstream applications may inhibit enzyme activity and result in impaired DNA cleavage. Certain sites in lambda DNA are difficult to cleave with Cfr42I, the same as with its prototype SacII. At least two copies of Cfr42I recognition site are required for efficient cleavage.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 10 U/μL
Incubator Temperature 37°C
Quantity 1200 U
Product Type Cfr42I (SacII)

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.