Learn More
CFP Mouse, anti-Human, CoraLite Plus 488, Clone: 1D5A9, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-66788-20UL
 View more versions of this product
Product Code. 18098175
  • £135.00 / 20µL
Description
The CFP antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CFP. This antibody recognizes human, pig, rat antigen. The CFP antibody has been validated for the following applications: WB analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
CFP
1D5A9
CoraLite Plus 488
Mouse
Primary
Human, Pig, Rat
155 kDa
IgG1
Western Blot
1000 μg/mL
BFD, CFP, Complement factor P, complement factor properdin, PFC, PFD, PROPERDIN
20 μL
5199
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.