missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CFLAR/FLIP Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | CFLAR/FLIP |
|---|---|
| Concentration | 0.72 mg/mL |
| Applications | Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16886753
|
Proteintech
10394-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16876753
|
Proteintech
10394-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FLIP (CFLAR) is an apoptosis regulator protein which functions as a crucial link between cell survival and cell death pathways in mammalian cells. It acts as an inhibitor of TNFRSF6 mediated apoptosis. A proteolytic fragment (p43) is likely retained in the death-inducing signaling complex (DISC) thereby blocking further recruitment and processing of caspase-8 at the complex. Full length and shorter isoforms have been shown either to induce apoptosis or to reduce TNFRSF-triggered apoptosis. Lacks enzymatic (caspase) activity.Specifications
| CFLAR/FLIP | |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human | |
| O15519 | |
| 8837 | |
| CFLAR/FLIP Fusion Protein Ag0593 | |
| Primary | |
| -20°C | |
| CFLAR |
| 0.72 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| c FLIP, c FLIPL, c FLIPR, c FLIPS, CASH, CASP8AP1, Caspase eight related protein, Caspase homolog, Casper, CFLAR, CLARP, FLAME, FLAME 1, FLAME1, FLIP, I FLICE, Inhibitor of FLICE, MRIT, Usurpin | |
| CFLAR | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title