missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CFHR5, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova H00081494-D01.100uL
This item is not returnable.
View return policy
Description
Sequence: MLLLFSVILISWVSTVGGEGTLCDFPKIHHGFLYDEEDYNPFSQVPTGEVFYYSCEYNFVSPSKSFWTRITCTEEGWSPTPKCLRMCSFPFVKNGHSESSGLIHLEGDTVQIICNTGYSLQNNEKNISCVERGWSTPPICSFTKGECHVPILEANVDAQPKKESYKVGDVLKFSCRKNLIRVGSDSVQCYQFGWSPNFPTCKGQVRSCGPPPQLSNGEVKEIRKEEYGHNEVVEYDCNPNFIINGPKKIQCVDGEWTTLPTCVEQVKTCGYIPELEYGYVQPSVPPYQHGVSVEVNCRNEYAMIGNNMITCINGIWTELPMCVATHQLKRCKIAGVNIKTLLKLSGKEFNHNSRIRYRCSDIFRYRHSVCINGKWNPEVDCTEKREQFCPPPPQIPNAQNMTTTVNYQDGEKVAVLCKENYLLPEAKEIVCKDGRWQSLPRCVESTAYCGPPPSINNGDTTSFPLSVYPPGSTVTYRCQSFYKLQGSVTVTCRNKQWSEPPRCLDPCVVSEENMNKNNIQLKWRNDGKLYAKTGDAVEFQCKFPHKAMISSPPFRAICQEGKFEYPICESpecifications
| CFHR5 | |
| Polyclonal | |
| No additive | |
| NM_030787 | |
| CFHR5 | |
| CFHR5 (NP_110414.1, 1 a.a. ∼ 569 a.a) full-length human protein. | |
| RUO | |
| 81494 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| complement factor H-related 5 | |
| CFHL5/FHR-5/FHR5/FLJ10549/MGC133240 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction