missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CFHR2 Mouse anti-Human, Clone: 6, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA530231
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human CFHR2.
Might be involved in complement regulation. Can associate with lipoproteins and may play a role in lipid metabolism.
Specifications
| CFHR2 | |
| Monoclonal | |
| Unconjugated | |
| CFHR2 | |
| CFHL2, FHR2, HFL3, DDESK59, FHR-2, H factor (complement)-like 3, complement factor H-related protein 2, factor H-related gene 2, h factor-like 3, h factor-like protein 2 | |
| Mouse | |
| Protein A | |
| RUO | |
| 3080 | |
| Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C | |
| Liquid |
| ELISA | |
| 6 | |
| PBS with no preservative | |
| P36980 | |
| CFHR2 | |
| Recombit Human CFHR2 Protein | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto