Learn More
CFD Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26050-1-AP-150UL
 View more versions of this product
Product Code. 16854575
  • £349.00 / 150µL
Description
Complement factor D is a serine protease of the alternative pathway of complement activation. The protein is a single chain polypeptide with a molecular mass of 25 kDa. Factor D cleaves factor B bound to C3b, generating the alternative pathway C3 convertase C3bBb and releasing the Ba fragment.
Specifications
Specifications
CFD
Polyclonal
Unconjugated
CFD
ADIPSIN, ADN, C3 convertase activator, CFD, Complement factor D, complement factor D (adipsin), DF, PFD, Properdin factor D
Rabbit
Antigen Affinity Chromatography
RUO
1675
-20°C
Liquid
Western Blot
0.23 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P00746
CFD
CFD Fusion Protein Ag23473
150 μL