missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CFC1B Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA548977

Product Code. 15633275

  • £441.00 / 400µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CFC1B Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CFC1B
Polyclonal
PBS with 0.09% sodium azide; pH 7.4
P0CG36
CFC1B
KLH conjugated synthetic peptide between 20-48 amino acids from the N-terminal region of human CFC1B.
400 μL
Primary
Human
Antibody
IgG
Western Blot
Unconjugated
CFC1B
CFC1B; cripto, FRL-1, cryptic family 1B; cryptic family protein 1B
Rabbit
Antigen affinity chromatography, Protein A
RUO
653275
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.