missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CFAP74 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-90919
This item is not returnable.
View return policy
Description
CFAP74 Polyclonal antibody specifically detects CFAP74 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| CFAP74 | |
| Polyclonal | |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| chromosome 1 open reading frame 222, Cilia And Flagella Associated Protein 74, Cilia- And Flagella-Associated Protein 74, KIAA1751, MGC133181 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:QELEAQKRAFEEEQKLRKQEIISRILKEEAEEEKRKKQHPPTSARHRLTLRDKTWNYISDFCKKTTVPTNTYTLDYEAAAGPGPSRLLEVVSSELIQ | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 85452 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction