missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CFAP61 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552649
This item is not returnable.
View return policy
Description
Immunogen sequence: HMKFNNLTLI STHGLPGKKL LDTEQRKFLA SDHCFNDKDY ALMSLCSWVN VVVGRMTGID RAAKHVVLST DEIVPYDHLI LCTGQQYQVP CPTEADISQH LTNREVPNSS QRRYTGKVPC NHFTLNEEED CFKALIWIRN NSITTE Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 17%.
Specifications
| CFAP61 | |
| Polyclonal | |
| Unconjugated | |
| CFAP61 | |
| 4930529M08Rik; BB178539; BC024760; C20orf26; CaM-IP3; CFAP61; cilia and flagella associated protein 61; cilia- and flagella-associated protein 61; dJ1002M8.3; dJ1178H5.4; RGD1311519 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 26074 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NHU2 | |
| CFAP61 | |
| Recombinant protein corresponding to Human CFAP61. Recombinant protein control fragment (Product #RP-90022). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction