missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CFAP161 Polyclonal antibody specifically detects CFAP161 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
Specifications
| Conjugate | mFluor Violet 500 SE |
| Dilution | Immunohistochemistry-Paraffin |
| Gene Alias | chromosome 15 open reading frame 26, FLJ38615, hypothetical protein LOC161502 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?