missing translation for 'onlineSavingsMsg'
Learn More

Cevimeline N,S-dioxide (Mixture of diastereomers), TRC

Product Code. 30357215
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30357215 50 mg 50mg
30354360 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30357215 Supplier Toronto Research Chemicals Supplier No. TRCC28352050MG
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C10 H17 N O3 S
Molecular Weight (g/mol) 231.312
Synonym (2'R,3R,3'S)-rel-2'-Methylspiro[1-azabicyclo[2.2.2]octane-3,5'-oxathiolane] 1,3'-Dioxide
IUPAC Name (2S,5S)-2-methyl-1'-oxidospiro[1,3-oxathiolane-5,3'-1-azoniabicyclo[2.2.2]octane] 3-oxide
SMILES C[C@H]1O[C@]2(CS1=O)C[N@+]3([O-])CC[C@@H]2CC3

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C10H17NO3S/c1-8-14-10(7-15(8)13)6-11(12)4-2-9(10)3-5-11/h8-9H,2-7H2,1H3/t8-,9-,10-,11+,15?/m0/s1
Formula Weight 231.093
Chemical Name or Material Cevimeline N,S-dioxide (Mixture of diastereomers)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.