missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cevimeline, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Description
Cevimeline (AF-102B) is a quinuclidine derivative of acetylcholine and a selective and orally active muscarinic M1 and M3 receptor agonist. Cevimeline stimulates secretion by the salivary glands and can be used as a sialogogue for xerostomia. Cevimeline can cross the blood-brain barrier (BBB).
Specifications
Specifications
| Chemical Name or Material | Cevimeline |
| Physical Form | Liquid |
| CAS | 107233-08-9 |
| Color | Brown |
| Quantity | 10 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C10H17NOS |
| For Use With (Application) | Neuroscience-Neuromodulation |
| Synonym | AF102B |
| SMILES | C[C@H]1SC[C@@]2(CN3CCC2CC3)O1 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction