missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cevimeline hydrochloride, Tocris Bioscience™
Selective M1 agonist
£193.30 - £681.41
Chemical Identifiers
CAS | 107220-28-0 |
---|---|
Molecular Formula | C10H18ClNOS |
Molecular Weight (g/mol) | 235.77 |
InChI Key | SURWTGAXEIEOGY-UHFFFAOYSA-N |
Synonym | hydrogen cevimeline chloride |
PubChem CID | 9837584 |
IUPAC Name | 2-methylspiro[1,3-oxathiolane-5,3'-1-azabicyclo[2.2.2]octane];hydrochloride |
SMILES | CC1OC2(CN3CCC2CC3)CS1.Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18732774
|
Tocris Bioscience
3689/10 |
10 mg |
£193.30
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18722574
|
Tocris Bioscience
3689/50 |
50 mg |
£681.41
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Selective M1 receptor agonist. Induces atropine-sensitive contractions of isolated guinea pig ilea and trachea preparations (EC50 values are 3.5 and 3 μM respectively). Reverses AF64A-induced cognitive impairments in vivo.Chemical Identifiers
107220-28-0 | |
235.77 | |
hydrogen cevimeline chloride | |
2-methylspiro[1,3-oxathiolane-5,3'-1-azabicyclo[2.2.2]octane];hydrochloride |
C10H18ClNOS | |
SURWTGAXEIEOGY-UHFFFAOYSA-N | |
9837584 | |
CC1OC2(CN3CCC2CC3)CS1.Cl |
Specifications
107220-28-0 | |
hydrogen cevimeline chloride | |
CC1OC2(CN3CCC2CC3)CS1.Cl | |
235.77 | |
235.77 | |
Cevimeline hydrochloride |
C10H18ClNOS | |
SURWTGAXEIEOGY-UHFFFAOYSA-N | |
2-methylspiro[1,3-oxathiolane-5,3'-1-azabicyclo[2.2.2]octane];hydrochloride | |
9837584 | |
>98% |
Safety and Handling
Recommended Storage : Store at -20°C