missing translation for 'onlineSavingsMsg'
Learn More

Cevimeline hydrochloride, MedChemExpress

Product Code. 16488168
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16488168 10 mg 10mg
16464077 50 mg 50mg
16410628 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16488168 Supplier Medchem Express Supplier No. HY70020B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cevimeline hydrochloride (AF102B hydrochloride) is a quinuclidine derivative of acetylcholine and a selective and orally active muscarinic M1 and M3 receptor agonist. Cevimeline hydrochloride stimulates secretion by the salivary glands and can be used as a sialogogue for xerostomia. Cevimeline hydrochloride can cross the blood-brain barrier (BBB).

Cevimeline hydrochloride (AF102B hydrochloride) is a quinuclidine derivative of acetylcholine and a selective and orally active muscarinic M1 and M3 receptor agonist. Cevimeline hydrochloride stimulates secretion by the salivary glands and can be used as a sialogogue for xerostomia. Cevimeline hydrochloride can cross the blood-brain barrier (BBB).

Specifications

Chemical Name or Material Cevimeline hydrochloride
Physical Form Solid
CAS 107220-28-0
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C10H18ClNOS
For Use With (Application) Neuroscience-Neuromodulation
Synonym AF102B hydrochloride
Solubility Information H2O : ≥ 50 mg/mL (212.07 mM)
SMILES C[C@H]1SC[C@@]2(CN3CCC2CC3)O1.Cl
Molecular Weight (g/mol) 235.77
Formula Weight 235.77
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.