missing translation for 'onlineSavingsMsg'
Learn More

Cevimeline hydrochloride hemihydrate, MedChemExpress

Product Code. 16444117
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16444117 50 mg 50mg
16468218 10 mg 10mg
16440658 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16444117 Supplier Medchem Express Supplier No. HY76772
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Cevimeline hydrochloride hemihydrate (SNI-2011) is a quinuclidine derivative of acetylcholine and a selective and orally active muscarinic M1 and M3 receptor agonist. Cevimeline hydrochloride hemihydrate stimulates secretion by the salivary glands and can be used as a sialogogue for xerostomia. Cevimeline hydrochloride hemihydrate can cross the blood-brain barrier (BBB).

Cevimeline hydrochloride hemihydrate (SNI-2011) is a quinuclidine derivative of acetylcholine and a selective and orally active muscarinic M1 and M3 receptor agonist. Cevimeline hydrochloride hemihydrate stimulates secretion by the salivary glands and can be used as a sialogogue for xerostomia. Cevimeline hydrochloride hemihydrate can cross the blood-brain barrier (BBB).

Specifications

Chemical Name or Material Cevimeline hydrochloride hemihydrate
Physical Form Solid
CAS 153504-70-2
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C10H17NOS.HCl.1/2H2O
For Use With (Application) Neuroscience-Neuromodulation
Synonym SNI-2011AF102B hydrochloride hemihydrate
Solubility Information H2O : ≥ 50 mg/mL (204.27 mM)
SMILES C[C@H]1SC[C@@]2(CN3CCC2CC3)O1.[H]Cl.[Relative Stereochemistry].[0.5 H2O]
Molecular Weight (g/mol) 244.78
Formula Weight 244.78
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.