missing translation for 'onlineSavingsMsg'
Learn More

Cevimeline hydrochloride hemihydrate, MedChemExpress

Product Code. 16440658
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16440658

Brand: Medchem Express HY76772

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cevimeline hydrochloride hemihydrate (SNI-2011) is a quinuclidine derivative of acetylcholine and a selective and orally active muscarinic M1 and M3 receptor agonist. Cevimeline hydrochloride hemihydrate stimulates secretion by the salivary glands and can be used as a sialogogue for xerostomia. Cevimeline hydrochloride hemihydrate can cross the blood-brain barrier (BBB).

Cevimeline hydrochloride hemihydrate (SNI-2011) is a quinuclidine derivative of acetylcholine and a selective and orally active muscarinic M1 and M3 receptor agonist. Cevimeline hydrochloride hemihydrate stimulates secretion by the salivary glands and can be used as a sialogogue for xerostomia. Cevimeline hydrochloride hemihydrate can cross the blood-brain barrier (BBB).

Specifications

Chemical Name or Material Cevimeline hydrochloride hemihydrate
Physical Form Solid
CAS 153504-70-2
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C10H17NOS.HCl.1/2H2O
For Use With (Application) Neuroscience-Neuromodulation
Synonym SNI-2011AF102B hydrochloride hemihydrate
Solubility Information H2O : ≥ 50 mg/mL (204.27 mM)
SMILES C[C@H]1SC[C@@]2(CN3CCC2CC3)O1.[H]Cl.[Relative Stereochemistry].[0.5 H2O]
Molecular Weight (g/mol) 244.78
Formula Weight 244.78
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.