missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cetuximab Recombinant Monoclonal Antibody
GREENER_CHOICE

Recombinant Monoclonal Antibody

£397.00 - £1002.00

Specifications

Antigen Cetuximab Chimeric
Concentration 1 mg/mL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Applications Functional Assay
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30283014
GREENER_CHOICE
View Documents
Invitrogen™
MA558305
100 μg
£397.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30282554
View Documents
Invitrogen™
MA558306
1 mg
£1002.00
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.

Cetuximab (Erbitux) is a chimeric (mouse/human) monoclonal antibody that is approved by the U.S. Food and Drug Administration for the treatment of patients with metastatic colorectal cancer, metastatic non-small cell lung cancer and head and neck cancer. Cetuximab, as an epidermal growth factor receptor (EGFR) inhibitor, binds to EGFR.
Specifications

Specifications

Cetuximab Chimeric
-20°C, Avoid Freeze/Thaw Cycles
Recombinant Monoclonal
Lyophilized
IgG1
Human
Erbitux; Fab C225; IMC-225
Antibody
1 mg/mL
Functional Assay
Unconjugated
Human
RUO
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Primary
Protein A
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.