missing translation for 'onlineSavingsMsg'
Learn More

Cetrimonium Chloride, TRC

Product Code. 18043420
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18043420 10 g 10g
18013920 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18043420 Supplier Toronto Research Chemicals Supplier No. TRCC28149510G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-02-7
Molecular Formula C19 H42 N . Cl
Molecular Weight (g/mol) 320
Synonym 1-Hexadecanaminium, N,N,N-trimethyl-, chloride (1:1), 1-Hexadecanaminium, N,N,N-trimethyl-, chloride (9CI), Ammonium, hexadecyltrimethyl-, chloride (8CI), Hexadecyltrimethylammonium chloride (6CI), 1631CL, Adogen 444, Adogen 444-29, Aliquat 1529, Aliquat 6, Ammonyx CETAC 25, Ammonyx Cetac, Ammonyx Cetac 30, Arquad 16, Arquad 16-25, Arquad 16-25LO, Arquad 16-25W, Arquad 16-26, Arquad 16-29, Arquad 16-29W, Arquad 16-50, BP 40, Barquat CT 29, C 16TAC, CA 2350, CETAC, CM 29, CTAC, CTAC 1, CTAC 30KC, CTAC CT-429, CTACL, CTMA, Carsoquat CT 425, Carsoquat CT 429, Catinal CTC 70ET, Catiogen TMP, Catiogen TMR, Cation PB 300, Cation PB 40, Cationic 1631, Cetac 30, Cetrimonium chloride, Cetyltrimethylammomium chloride, Cetyltrimethylammonium chloride, D 1631, Dehyquart A, Dehyquart A-CA, Dehyquart A-CE, Dodigen 1383, FSM 28, Genamin CTAC, Genamin CTAC 50, Genamin CTAC 50ET, HDTMA-Cl, HTAC, Incroquat CTC 30, Intexan CTC 29, Intexsan CTC 29, Intexsan CTC 50, Lebon TM 16, Lebon TM 60, Morpan CHA, N,N,N-Trimethyl-1-hexadecanaminium chloride, N-Palmityl-N,N,N-trimethylammonium chloride, Nissan Cation PB 40, Nissan Cation PB 40R, PB 40, Palmityltrimethylammonium chloride, Pionin B 611, Protaquat CT 29, Quartamin 60W, Quartamin 60W30, Quatramine C 16/29, Saboquat, Surfroyal CTAC, Swanol CA 2350, Trimethylcetylammonium chloride, Trimethylhexadecylammonium chloride, Variquat E 228, Varisoft 300, n-Hexadecyltrimethylammonium chloride
IUPAC Name hexadecyl(trimethyl)azanium;chloride
SMILES [Cl-].CCCCCCCCCCCCCCCC[N+](C)(C)C

Specifications

Quantity 10 g
InChI Formula InChI=1S/C19H42N.ClH/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(2,3)4;/h5-19H2,1-4H3;1H/q+1;/p-1
Formula Weight 319.3006
Chemical Name or Material Hexadecyltrimethylammonium chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.