missing translation for 'onlineSavingsMsg'
Learn More

Cetrimonium bromide, MedChemExpress

Product Code. 16478706
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16478706 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16478706 Supplier Medchem Express Supplier No. HYB1260
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Cetrimonium bromide (CTAB) is an amine based cationic quaternary surfactant, is one of the components of the topical antiseptic Cetrimide.

Cetrimonium bromide (CTAB) is an amine based cationic quaternary surfactant, is one of the components of the topical antiseptic Cetrimide.

Specifications

Chemical Name or Material Cetrimonium bromide
CAS 57-09-0
Color White
Purity Grade Notes Research
Molecular Formula C19H42BrN
Quantity 100 mg
Synonym CTABCetyltrimethylammonium bromideHexadecyltrimethylammonium bromide
Solubility Information DMSO : 10 mg/mL (27.44 mM; Need ultrasonic) ∣H2O : 4.55 mg/mL (12.48 mM; Need ultrasonic)
SMILES CCCCCCCCCCCCCCCC[N+](C)(C)C.[Br-]
Molecular Weight (g/mol) 364.45
Formula Weight 364.45
Percent Purity 98.0%
Grade Research
Physical Form Powder
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.