missing translation for 'onlineSavingsMsg'
Learn More

Cetirizine N-Oxide, Mikromol™

Product Code. 17913776
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity unitSize
17913776 100 mg 100mg
1 options
This item is not returnable. View return policy

Produktkod. 17913776

Brand: Mikromol MM0380.13

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Cetirizine N-Oxide
TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material Cetirizine N-Oxide
CAS 1076199-80-8
Analyte or Component Names Cetirizine N-Oxide
Shipping Condition Room Temperature
Molecular Formula C21 H25 Cl N2 O4
Certifications/Compliance ISO 9001
Synonym Cetirizine N-Oxide, 2-[2-[4-[(4-Chlorophenyl)phenylmethyl]-1-oxido-1-piperazinyl]ethoxy]acetic acid
InChI Formula InChI=1S/C21H25ClN2O4/c22-19-8-6-18(7-9-19)21(17-4-2-1-3-5-17)23-10-12-24(27,13-11-23)14-15-28-16-20(25)26/h1-9,21H,10-16H2,(H,25,26)
SMILES OC(=O)COCC[N+]1([O-])CCN(CC1)C(c2ccccc2)c3ccc(Cl)cc3
IUPAC Name 2-[2-[4-[(4-chlorophenyl)-phenylmethyl]-1-oxidopiperazin-1-ium-1-yl]ethoxy]acetic acid
Molecular Weight (g/mol) 404.89
Quantity 100 mg
Formula Weight 404.1503 g/mol
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.