missing translation for 'onlineSavingsMsg'
Learn More

Cetirizine dihydrochloride, MedChemExpress

Product Code. 16457246
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16457246 100 mg 100mg
16421836 500 mg 500mg
16411246 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16457246 Supplier Medchem Express Supplier No. HY17042A
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Cetirizine dihydrochloride, a second-generation antihistamine and the carboxylated metabolite of hydroxyzine, is a specific, orally active and long-acting histamine H1-receptor antagonist. Cetirizine dihydrochloride marks antiallergic properties and inhibits eosinophil chemotaxis during the allergic response.

Cetirizine dihydrochloride, a second-generation antihistamine and the carboxylated metabolite of hydroxyzine, is a specific, orally active and long-acting histamine H1-receptor antagonist. Cetirizine dihydrochloride marks antiallergic properties and inhibits eosinophil chemotaxis during the allergic response.

Specifications

Chemical Name or Material Cetirizine dihydrochloride
Physical Form Solid
CAS 83881-52-1
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C21H27Cl3N2O3
For Use With (Application) COVID-19-immunoregulation
Synonym P071
Solubility Information DMSO : 100 mg/mL (216.54 mM; Need ultrasonic) ∣H2O : 100 mg/mL (216.54 mM; Need ultrasonic)
SMILES O=C(O)COCCN1CCN(C(C2=CC=C(Cl)C=C2)C3=CC=CC=C3)CC1.Cl.Cl
Molecular Weight (g/mol) 461.81
Formula Weight 461.81
Percent Purity 99.17%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.