missing translation for 'onlineSavingsMsg'
Learn More

Cesium titanium oxide, 99.85% (metals basis)

Product Code. 11373598
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11373598 25 g 25g
11383598 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11373598 Supplier Thermo Scientific Alfa Aesar Supplier No. 039207.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12158-57-5

It is employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12158-57-5
MDL Number MFCD00053969
Synonym Cesium titanate

Specifications

Color White
Physical Form Powder
Quantity 25 g
Linear Formula Cs2TiO3
Solubility Information Insoluble in water.
Formula Weight 361.71
Percent Purity 99.85%
Assay (metals basis)
Chemical Name or Material Cesium titanium oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.