missing translation for 'onlineSavingsMsg'
Learn More

Cesium Sulfate, puriss. p.a., ≥99.5% (T), Honeywell™ Fluka™

Product Code. 15611920 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
50 g
Packaging:
Glass Bottle
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15611920 50 g Glass Bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15611920 Supplier Honeywell-Fluka Supplier No. 2101950G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 1019°C
CAS 10294-54-9
Color White
pH 6.0 to 7.5 (at 723.7g/L 25°C)
Quantity 50 g
Molecular Formula Cs2O4S
MDL Number MFCD00010959
UN Number NONH for all modes of transport
Synonym cesium sulfate, dicesium sulfate, sulfuric acid, dicesium salt, caesium sulphate, caesium sulfate, unii-8d6r91cs62, dicaesium 1+ ion sulfate, cesiumsulfate, dicaesium 1+ sulfate
Solubility Information Soluble in water, 0.2g/mL
InChI Key FLJPGEWQYJVDPF-UHFFFAOYSA-L
SMILES [Cs+].[Cs+].[O-]S([O-])(=O)=O
IUPAC Name dicaesium(1+) sulfate
Molecular Weight (g/mol) 361.87
PubChem CID 25137
Formula Weight 361.87g/mol
Percent Purity ≥99.5% (T)
Grade puriss. p.a.
Packaging Glass Bottle
Appearance Crystals
Density 4.243g/mL at 25°C
Chemical Name or Material Cesium Sulfate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.