missing translation for 'onlineSavingsMsg'
Learn More

Cesium sulfate, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals

Catalog Number 11386128
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
10294-54-9
Cs2O4S
361.87
MFCD00010959
FLJPGEWQYJVDPF-UHFFFAOYSA-L
cesium sulfate, dicesium sulfate, sulfuric acid, dicesium salt, caesium sulphate, caesium sulfate, unii-8d6r91cs62, dicaesium 1+ ion sulfate, cesiumsulfate, dicaesium 1+ sulfate
25137
dicesium;sulfate
[Cs+].[Cs+].[O-]S([O-])(=O)=O
This item is not returnable. View return policy
10294-54-9
Cs2O4S
361.87
MFCD00010959
FLJPGEWQYJVDPF-UHFFFAOYSA-L
cesium sulfate, dicesium sulfate, sulfuric acid, dicesium salt, caesium sulphate, caesium sulfate, unii-8d6r91cs62, dicaesium 1+ ion sulfate, cesiumsulfate, dicaesium 1+ sulfate
25137
dicesium;sulfate
[Cs+].[Cs+].[O-]S([O-])(=O)=O

Cesium sulfate is used to prepare dense aqueous solutions for use in isopycnic centrifugation. It is also used for crystal growing purposes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 1,010°C
Color White
Physical Form Powder
Quantity 10 g
Linear Formula Cs2SO4
Merck Index 14,2016
Solubility Information Soluble in water (1820g/L).
Formula Weight 361.87
Percent Purity 99.997%
Grade Puratronic™
Odor Odorless
Sensitivity Hygroscopic
Density 4.243 g/mL
Assay (metals basis)
Chemical Name or Material Cesium sulfate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'