missing translation for 'onlineSavingsMsg'
Learn More

Cesium sulfate, 99%

Product Code. 11422788
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11422788 500 g 500g
11402788 25 g 25g
11412788 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11422788 Supplier Thermo Scientific Alfa Aesar Supplier No. A16767.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10294-54-9 | Cs2O4S | 361.87 g/mol

Cesium sulfate is used to prepare dense aqueous solutions for use in isopycnic centrifugation. It is used as metal oxide catalyst enhancer for heterogeneous processes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cesium sulfate is used to prepare dense aqueous solutions for use in isopycnic centrifugation. It is used as metal oxide catalyst enhancer for heterogeneous processes.

Solubility
Soluble in water. (1820 g/L) at 20°C

Notes
Hygroscopic. Protect from humidity and water. Store under dry inert gas. Incompatible with water, reducing agents and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10294-54-9
Molecular Formula Cs2O4S
Molecular Weight (g/mol) 361.87
MDL Number MFCD00010959
InChI Key FLJPGEWQYJVDPF-UHFFFAOYSA-L
Synonym cesium sulfate, dicesium sulfate, sulfuric acid, dicesium salt, caesium sulphate, caesium sulfate, unii-8d6r91cs62, dicaesium 1+ ion sulfate, cesiumsulfate, dicaesium 1+ sulfate
PubChem CID 25137
IUPAC Name dicesium;sulfate
SMILES [Cs+].[Cs+].[O-]S([O-])(=O)=O

Specifications

Melting Point 1,010°C
Quantity 500 g
Merck Index 14,2016
Solubility Information Soluble in water. (1820g/L) at 20°C
Formula Weight 361.87
Percent Purity 99%
Odor Odorless
Sensitivity Hygroscopic
Density 4.24
Chemical Name or Material Cesium sulfate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.