missing translation for 'onlineSavingsMsg'
Learn More

Cesium Sulfate, ≥99%, Grade I, Honeywell Fluka™

Product Code. 15677740 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Poly bottle
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15677740 50 g Poly bottle 50g
15667740 250 g Poly bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15677740 Supplier Honeywell-Fluka Supplier No. C313650G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Grade I,≥99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10294-54-9
Molecular Formula Cs2O4S
Molecular Weight (g/mol) 361.87
MDL Number MFCD00010959
InChI Key FLJPGEWQYJVDPF-UHFFFAOYSA-L
Synonym cesium sulfate, dicesium sulfate, sulfuric acid, dicesium salt, caesium sulphate, caesium sulfate, unii-8d6r91cs62, dicaesium 1+ ion sulfate, cesiumsulfate, dicaesium 1+ sulfate
PubChem CID 25137
IUPAC Name dicaesium(1+) sulfate
SMILES [Cs+].[Cs+].[O-]S([O-])(=O)=O

Specifications

Color White
pH 6.0 to 7.5 (at 723.7g/L 25°C)
Physical Form Crystalline
Quantity 50 g
UN Number NONH for all modes of transport
Formula Weight 361.87g/mol
Percent Purity ≥99%
Packaging Poly bottle
Density 4.243g/mL at 25°C
Chemical Name or Material Cesium sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.