missing translation for 'onlineSavingsMsg'
Learn More

Cesium silicate (meta), 99%, Thermo Scientific Chemicals

Product Code. 11329357
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11329357 25 g 25g
11319357 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11329357 Supplier Thermo Scientific Alfa Aesar Supplier No. 040208.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15586-77-3
Molecular Formula Cs2O3Si
Molecular Weight (g/mol) 341.89
MDL Number MFCD00798519
InChI Key MZRJYMUQLBFTMG-UHFFFAOYSA-N
Synonym dicaesium oxosilanebis olate, caesium silicate, dicaesium 1+ ion oxosilanebis olate, cesium metasilicate trace metals basis 100g
PubChem CID 12893658
IUPAC Name dicesium;dioxido(oxo)silane
SMILES [Cs+].[Cs+].[O-][Si]([O-])=O

Specifications

Physical Form Powder
Quantity 25 g
Solubility Information Insoluble in water.
Formula Weight 341.89
Percent Purity 99%
Odor Odorless
Chemical Name or Material Cesium silicate (meta)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.