missing translation for 'onlineSavingsMsg'
Learn More

Cesium propionate, complex with propionic acid, Thermo Scientific Chemicals

Product Code. 11343507
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11343507 50 g 50g
11353507 2 g 2g
11363507 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11343507 Supplier Thermo Scientific Alfa Aesar Supplier No. 018865.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38869-24-8
Molecular Formula C3H5CsO2
Molecular Weight (g/mol) 205.98
MDL Number MFCD00014025
InChI Key YBZSHUAKOJGWRT-UHFFFAOYSA-M
Synonym cesium propionate, caesium propanoate, cesium propionate, compound with propionic acid, acmc-20alh6, propionic acid cesium salt, caesium 1+ propionate, caesium 1+ ion propionate
PubChem CID 23693496
IUPAC Name cesium;propanoate
SMILES [Cs+].CCC([O-])=O

Specifications

Quantity 50 g
Sensitivity Hygroscopic
Solubility Information Soluble in dimethyl formamide,dimethyl sulfoxide.
Formula Weight 280.06
Physical Form Crystalline
Chemical Name or Material Cesium propionate, Complex with propionic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.