missing translation for 'onlineSavingsMsg'
Learn More

Cesium, plasma standard solution, Specpure™ Cs 1000μg/mL

Product Code. 11391369
Change view
Click to view available options
Quantity:
50 mL
500 mL
Unit Size:
500mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11391369 50 mL 50mL
11410340 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11391369 Supplier Thermo Scientific Alfa Aesar Supplier No. 013825.AD
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 534-17-8 | CCs2O3 | 325.82 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 534-17-8 , 7697-37-2 , 7732-18-5
Molecular Formula CCs2O3
Molecular Weight (g/mol) 325.82
MDL Number MFCD00010957
InChI Key FJDQFPXHSGXQBY-UHFFFAOYSA-L
PubChem CID 10796
SMILES [Cs+].[Cs+].[O-]C([O-])=O

Specifications

Vapor Pressure 23 hPa (17mm Hg) at 20°C
Physical Form Liquid
Color Colorless
IUPAC Name dicesium;carbonate
Quantity 50 mL
Concentration or Composition (by Analyte or Components) Nitric acid: 5.0%; Cesium carbonate: 0.122%; Water: 94.878%
Chemical Name or Material Cesium

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.