missing translation for 'onlineSavingsMsg'
Learn More

Cesium perchlorate, Reagent Grade, Thermo Scientific Chemicals

Product Code. 11341669
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11341669 25 g 25g
11331669 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11341669 Supplier Thermo Scientific Alfa Aesar Supplier No. 012308.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cesium perchlorate is used for long-duration optical transmission of information with infofuses. It is a modifier of polymer electrolyte/luminescent conjugated polymer mixed ionic-electronic conductors. It has been used as a reactant for investigating metal salt effects on morphology of silica nanomaterials prepared by polymerization on hydrogels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13454-84-7
Molecular Formula ClCsO4
Molecular Weight (g/mol) 232.351
MDL Number MFCD00010956
InChI Key WKDKOOITVYKILI-UHFFFAOYSA-M
Synonym cesium perchlorate, caesium 1+ ion perchlorate, perchloric acid, cesium salt, caesiumperchlorat, caesium perchlorate, perchloric acid, cesium salt 1:1, clo4.cs, acmc-20alh8, caesium 1+ perchlorate ion
PubChem CID 123309
IUPAC Name cesium;perchlorate
SMILES [O-]Cl(=O)(=O)=O.[Cs+]

Specifications

Melting Point 250°C (decomposition)
Physical Form Powder
Quantity 25 g
UN Number UN1481
Solubility Information Soluble in water.
Formula Weight 232.35
Grade Reagent
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Cesium perchlorate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.