missing translation for 'onlineSavingsMsg'
Learn More

Cesium perchlorate, Reagent Grade, Thermo Scientific Chemicals

Product Code. p-7048973
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11341669

Brand: Thermo Scientific Alfa Aesar 012308.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cesium perchlorate is used for long-duration optical transmission of information with infofuses. It is a modifier of polymer electrolyte/luminescent conjugated polymer mixed ionic-electronic conductors. It has been used as a reactant for investigating metal salt effects on morphology of silica nanomaterials prepared by polymerization on hydrogels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13454-84-7
Molecular Formula ClCsO4
Molecular Weight (g/mol) 232.351
MDL Number MFCD00010956
InChI Key WKDKOOITVYKILI-UHFFFAOYSA-M
Synonym cesium perchlorate, caesium 1+ ion perchlorate, perchloric acid, cesium salt, caesiumperchlorat, caesium perchlorate, perchloric acid, cesium salt 1:1, clo4.cs, acmc-20alh8, caesium 1+ perchlorate ion
PubChem CID 123309
IUPAC Name cesium;perchlorate
SMILES [O-]Cl(=O)(=O)=O.[Cs+]

Specifications

Melting Point 250°C (decomposition)
Physical Form Powder
Quantity 25 g
UN Number UN1481
Solubility Information Soluble in water.
Formula Weight 232.35
Grade Reagent
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Cesium perchlorate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.