missing translation for 'onlineSavingsMsg'
Learn More

Cesium orthovanadate, 99.9% (metals basis)

Product Code. 11393598
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11393598 10 g 10g
11303608 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11393598 Supplier Thermo Scientific Alfa Aesar Supplier No. 039211.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 34283-69-7

Cesium orthovanadate, is widely used in chemical research, and also as an intermediate in Pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34283-69-7
MDL Number MFCD00210609
Synonym cesium orthovanadate, tricaesium 1+ vanadate 3-

Specifications

Physical Form Powder
Quantity 10 g
UN Number UN3288
Solubility Information It is soluble in water.
Formula Weight 513.65
Percent Purity 99.9%
Assay (metals basis)
Chemical Name or Material Cesium orthovanadate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.