missing translation for 'onlineSavingsMsg'
Learn More

Cesium nitrate, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11378937
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Cesium nitrate prisms are used in infrared spectroscopy, x-ray phosphors and scintillation counters. It is a precursor material for the synthesis of cesium nitratocuprate and cesium salts. Cesium nitrate doped on carbon-based ruthenium catalyst plays an important role in synthesis of ammonia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-18-6
Molecular Formula CsNO3
Molecular Weight (g/mol) 194.909
MDL Number MFCD00010963
InChI Key NLSCHDZTHVNDCP-UHFFFAOYSA-N
Synonym cesium nitrate, caesium nitrate, nitric acid, cesium salt, cesium i nitrate 1:1, caesium 1+ nitronate, acmc-1bhhg, caesium 1+ ion nitronate, ksc492a7j
PubChem CID 62674
IUPAC Name cesium;nitrate
SMILES [N+](=O)([O-])[O-].[Cs+]

Specifications

Boiling Point (decomposition)
Melting Point 414°C
Color White
Physical Form Crystalline
Quantity 100 g
Assay Percent Range 99.999% (Metal basis)
Solubility Information Soluble in water and acetone. Slightly soluble in ethanol.
Formula Weight 194.91
Odor Odorless
Refractive Index 1.55
Sensitivity Hygroscopic
Appearance White crystalline powder
Density 3.68 g/cm3 at 20°C
Chemical Name or Material Cesium nitrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'