missing translation for 'onlineSavingsMsg'
Learn More

Cesium nitrate, 99.9%, (trace metal basis), for analysis, Thermo Scientific Chemicals

Product Code. 13458438
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-18-6
Molecular Formula CsNO3
Molecular Weight (g/mol) 194.91
MDL Number MFCD00010963
InChI Key NLSCHDZTHVNDCP-UHFFFAOYSA-N
Synonym cesium nitrate, caesium nitrate, nitric acid, cesium salt, cesium i nitrate 1:1, caesium 1+ nitronate, acmc-1bhhg, caesium 1+ ion nitronate, ksc492a7j
PubChem CID 62674
IUPAC Name cesium;nitrate
SMILES [N+](=O)([O-])[O-].[Cs+]

Specifications

Melting Point 414.0°C
Color White
Physical Form Crystalline Powder and/or Crystals
Quantity 1 kg
Assay Percent Range 99.9%
Linear Formula CsNO3
UN Number 1451
Merck Index 15,2014
Solubility Information Solubility in water: 9.16 g/100mL (0°C). Other solubilities: soluble in acetone, slightly soluble in alcohol
Formula Weight 194.91
Percent Purity 99.9%
Grade Trace Metal Basis
Packaging Glass Bottle
Specific Gravity 3.68
Density 3.6800g/mL
Chemical Name or Material Cesium Nitrate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cesium nitrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Oxidising solid Category 1
  • Reproductive toxicity Category 2
Hazard Statement
  • H361-Suspected of damaging fertility or the unborn child.
  • H302-Harmful if swallowed.
  • H271-May cause fire or explosion; strong oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'