missing translation for 'onlineSavingsMsg'
Learn More

Cesium nitrate, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog Number 11302398
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
This item is not returnable. View return policy
7789-18-6
CsNO3
194.909
MFCD00010963
NLSCHDZTHVNDCP-UHFFFAOYSA-N
cesium nitrate, caesium nitrate, nitric acid, cesium salt, cesium i nitrate 1:1, caesium 1+ nitronate, acmc-1bhhg, caesium 1+ ion nitronate, ksc492a7j
62674
cesium;nitrate
[N+](=O)([O-])[O-].[Cs+]
This item is not returnable. View return policy
7789-18-6
CsNO3
194.909
MFCD00010963
NLSCHDZTHVNDCP-UHFFFAOYSA-N
cesium nitrate, caesium nitrate, nitric acid, cesium salt, cesium i nitrate 1:1, caesium 1+ nitronate, acmc-1bhhg, caesium 1+ ion nitronate, ksc492a7j
62674
cesium;nitrate
[N+](=O)([O-])[O-].[Cs+]

Cesium nitrate prisms are used in infrared spectroscopy, x-ray phosphors and scintillation counters. It is a precursor material for the synthesis of cesium nitratocuprate and cesium salts. Cesium nitrate doped on carbon-based ruthenium catalyst plays an important role in synthesis of ammonia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point (decomposition)
Melting Point 414°C
Color White
Physical Form Crystalline
Quantity 500 g
UN Number UN1451
Merck Index 14,2015
Solubility Information Soluble in water and acetone. Slightly soluble in ethanol.
Formula Weight 194.91
Percent Purity 99.99%
Odor Odorless
Refractive Index 1.55
Sensitivity Hygroscopic
Density 3.68
Assay (metals basis)
Chemical Name or Material Cesium nitrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'