missing translation for 'onlineSavingsMsg'
Learn More

Cesium Iodide, TraceSELECT™, ≥99.9995%, Solstice

Product Code. 15641560 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15641560 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15641560 Supplier Honeywell-Fluka Supplier No. 1672225G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

TraceSELECT™,≥99.9995%

TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions

 


 

TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 1280°C
Melting Point 626°C
CAS 7789-17-5
Color White
Physical Form Solid
Quantity 25 g
Molecular Formula CsI
MDL Number MFCD00010962
UN Number NONH for all modes of transport
Synonym cesium iodide, caesium iodide, cesium monoiodide, dicesium diiodide, tricesium triiodide, cesiumiodide, cesium iodide cs2i2, cesium iodide cs3i3, unii-u1p3gvc56l
InChI Key XQPRBTXUXXVTKB-UHFFFAOYSA-M
SMILES [I-].[Cs+]
IUPAC Name cesium;iodide
Molecular Weight (g/mol) 259.81
PubChem CID 24601
Formula Weight 259.81g/mol
Percent Purity ≥99.9995%
Grade TraceSELECT
Packaging Glass bottle
Density 4.51g/mL at 25°C
Chemical Name or Material Cesium Iodide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.