missing translation for 'onlineSavingsMsg'
Learn More

Cesium iodide, 99.999% (metals basis)

Product Code. 11371588
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11371588 100 g 100g
11381588 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11371588 Supplier Thermo Scientific Alfa Aesar Supplier No. 010992.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7789-17-5 | CsI | 259.81 g/mol

Cesium iodide is used as an input phosphor in x-ray image intensifier tube of fluoroscopy equipment. It is also used as an equipment for electromagnetic calorimetry in experimental particle physics. It is often employed as a beamsplitter in Fourier Transform Infrared (FT-IR) spectrometers. Cesium iodide is an inorganic scintillator which is mainly used in experimental nuclear physics, high energy physics, nuclear medicine, nuclear tomography, environmental studies and many other fields of use. It is utilized in calorimeters and detectors and also employed as a phosphor in fluoroscopy equipments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-17-5
Molecular Formula CsI
Molecular Weight (g/mol) 259.81
MDL Number MFCD00010962
InChI Key XQPRBTXUXXVTKB-UHFFFAOYSA-M
Synonym cesium iodide, caesium iodide, cesium monoiodide, dicesium diiodide, tricesium triiodide, cesiumiodide, cesium iodide cs2i2, cesium iodide cs3i3, unii-u1p3gvc56l
PubChem CID 24601
IUPAC Name cesium;iodide
SMILES [I-].[Cs+]

Specifications

Boiling Point 1280°C
Melting Point 626°C
Color White
Physical Form Crystalline
Quantity 100 g
Merck Index 14,2014
Solubility Information Soluble in water. Soluble in water and ethanol. Slightly soluble in methanol. Insoluble in acetone.
Formula Weight 259.81
Percent Purity 99.999%
Odor Odorless
Refractive Index 1.7876
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Cesium iodide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.