missing translation for 'onlineSavingsMsg'
Learn More

Cesium hydroxide monohydrate, 96%; cesium carbonate <5%

Product Code. 11470657
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11470657 25 g 25g
11334569 5 g 5g
11480657 100 g 100g
11490657 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11470657 Supplier Thermo Scientific Alfa Aesar Supplier No. B22480.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 35103-79-8 | CsH3O2 | 167.93 g/mol

Cesium hydroxide monohydrate can be used as a strong electrolyte for the battery and it also can be used as polymerization catalyst. It is also used as a raw materials for manufacturing various cesium salts and Catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cesium hydroxide monohydrate can be used as a strong electrolyte for the battery and it also can be used as polymerization catalyst. It is also used as a raw materials for manufacturing various cesium salts and Catalyst.

Solubility
Soluble in water with exothermic reaction.

Notes
Air sensitive & hygroscopic. Incompatible with oxidizing agents, acids, air, water/moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35103-79-8
Molecular Formula CsH3O2
Molecular Weight (g/mol) 167.93
MDL Number MFCD00149664
InChI Key ABSOMGPQFXJESQ-UHFFFAOYSA-M
Synonym cesium hydroxide monohydrate, cesium hydroxide hydrate, cesium hydroxide, monohydrate, cesium hydroxide-hydrate, cesium hydroxide, hydrate, ksc493c5l, caesium 1+ ion hydrate hydroxide, cesium hydroxide,monohydrate 9ci
PubChem CID 23679066
SMILES O.[OH-].[Cs+]

Specifications

Melting Point 205°C to 206°C
Quantity 25 g
UN Number UN3262
Merck Index 14,2013
Solubility Information Soluble in water with exothermic reaction.
IUPAC Name caesium(1+) hydrate hydroxide
Formula Weight 167.93 (149.91 Anhydrous)
Percent Purity 96%
Odor Odorless
Sensitivity Air sensitive; Hygroscopic
Density 1.87
Assay cesium carbonate <5%
Chemical Name or Material Cesium hydroxide monohydrate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cesium hydroxide monohydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H361f-Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Cesium hydroxide monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.