missing translation for 'onlineSavingsMsg'
Learn More

Cesium hydroxide, 50% w/w aq. soln., 99.9% (metals basis)

Product Code. 11365729
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11365729 25 g 25g
11375729 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11365729 Supplier Thermo Scientific Alfa Aesar Supplier No. 010020.ND
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 21351-79-1 | CsHO | 149.91 g/mol

Cesium hydroxide is used in electric storage batteries. Also used to make porous cesium titanosilicates and potential ion-exchange materials for cleanup of radioactive wastes. Cesium Hydroxide is used as electrolyte in alkaline batteries at subzero temperatures, color photography and as a catalyst in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21351-79-1
Molecular Formula CsHO
Molecular Weight (g/mol) 149.91
MDL Number MFCD00010964
InChI Key HUCVOHYBFXVBRW-UHFFFAOYSA-M
Synonym cesium hydroxide, caesium hydroxide, cesium hydrate, csoh, cesium hydroxide cs oh, cesium hydroxide dimer, cesium hydoxide, cs.ho, caesium 1+ oh, cesium hydroxide solution
PubChem CID 62750
ChEBI CHEBI:33988
IUPAC Name cesium;hydroxide
SMILES [OH-].[Cs+]

Specifications

Color Colorless
Physical Form Liquid
Quantity 25 g
UN Number UN2681
Solubility Information Highly soluble in water.
Formula Weight 149.91
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) 50% w/w aq. soln.
Odor Odorless
Sensitivity Air Sensitive
Density 1.725 g/mL
Assay (metals basis)
Chemical Name or Material Cesium hydroxide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cesium hydroxide, 50% w/w Aqueous Solution
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to accordance with local/regional/national/international regulations..
Supplemental information
  • MIXTURE LIST-Contain: Cesium hydroxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.