missing translation for 'onlineSavingsMsg'
Learn More

Cesium formate, 98%

Product Code. 11319719
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11319719

Brand: Thermo Scientific Alfa Aesar B21327.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3495-36-1 | CH2CsO2 | 178.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3495-36-1
Molecular Formula CH2CsO2
Molecular Weight (g/mol) 178.93
MDL Number MFCD00149883
InChI Key ATZQZZAXOPPAAQ-UHFFFAOYSA-N
Synonym cesium formate, formic acid, cesium salt, formic acid, cesium salt 1:1, formic acid cesium salt, 64-18-6 parent, cesuim formate, acmc-1ajhs, cho2.cs, caesium 1+ formate
PubChem CID 23663999
IUPAC Name caesium(1+) formic acid
SMILES [Cs+].OC=O

Specifications

Quantity 100 g
Beilstein 3912426
Formula Weight 177.92
Percent Purity 98%
Sensitivity Hygroscopic
Chemical Name or Material Cesium formate
compliance-icons
Product Identifier
  • Cesium formate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Formic acid, cesiumsalt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.