missing translation for 'onlineSavingsMsg'
Learn More

Cesium fluoride, 99% (metals basis)

Product Code. 11317617
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11317617 100 g 100g
11307617 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11317617 Supplier Thermo Scientific Alfa Aesar Supplier No. 012885.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13400-13-0 | CsF | 151.90 g/mol

Used for desilylation reactions in organic synthesis. Useful base in organic chemistry, due to its low nucleophilicity. Owing to its high ionic character, it is a very reactive source of fluoride ion in organic synthesis. Reagent of choice when reactive naked fluoride is required. Effective catalyst in promoting Knoevenegal condensation. Single crystals of CsF are useful as the windows of cells for infrared spectroscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13400-13-0
Molecular Formula CsF
Molecular Weight (g/mol) 151.90
MDL Number MFCD00010960
InChI Key XJHCXCQVJFPJIK-UHFFFAOYSA-M
Synonym cesium fluoride, caesium fluoride, cesium monofluoride, dicesium difluoride, tricesium trifluoride, unii-t76a371hjr, caesium 1+ ion fluoride, caesiumfluoride, cesiumfluoride
PubChem CID 25953
SMILES [F-].[Cs+]

Specifications

Boiling Point 1251°C
Melting Point 682°C
Color White
Physical Form Crystalline
Quantity 100 g
Assay Percent Range 99% (Metals basis)
UN Number UN2923
Solubility Information Soluble in water. Insoluble in dioxane and pyridine.
IUPAC Name caesium(1+) fluoride
Formula Weight 151.9
Percent Purity 99%
Odor Odorless
Sensitivity Hygroscopic
Density 4.115
Assay (metals basis)
Chemical Name or Material Cesium fluoride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.