missing translation for 'onlineSavingsMsg'
Learn More

Cesium chromate, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11396858
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Cesium chromate is used to produce cesium vapor by reaction with silicon, boron, or titanium, which is used in the final stages of creating vacuum tubes. It is used in the production of photo-tubes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13454-78-9
Molecular Formula CrCs2O4
Molecular Weight (g/mol) 381.80
MDL Number MFCD00016050
InChI Key BROHICCPQMHYFY-UHFFFAOYSA-N
Synonym cesium chromate, cesium chromate 99.9+%, dicesium dioxido dioxo chromium, dicesium chromate, cesium chromate cs2cro4, chromic acid h2cro4 , dicesium salt, caesium chromate, cro4.2cs, acmc-1bym2, dicaesium 1+ chromate 2-
PubChem CID 61613
IUPAC Name dicesium;dioxido(dioxo)chromium
SMILES [Cs+].[Cs+].[O-][Cr]([O-])(=O)=O

Specifications

Physical Form Powder
Quantity 25 g
UN Number UN3087
Solubility Information Soluble in water.
Formula Weight 381.81
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Cesium chromate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'